Merck
  • Mucoadhesive beads of gellan gum/pectin intended to controlled delivery of drugs.
Mucoadhesive beads of gellan gum/pectin intended to controlled delivery of drugs.
Carbohydrate polymers (2014-09-27)
Fabíola Garavello Prezotti, Beatriz Stringhetti Ferreira Cury, Raul Cesar Evangelista
ABSTRACT
Gellan gum/pectin beads were prepared by ionotropic gelation, using Al(3+) as crosslinker. High yield (92.76%) and entrapment efficiency (52.22-88.78%) were reached. Beads exhibited high circularity (0.730-0.849) and size between 728.95 and 924.56 μm. Particle size and circularity was increased by raising polymer and crosslinker concentrations. Polymers ratio did not influence beads properties. The materials stability and the absence of drug-polymers interactions were evidenced by thermal analysis and FTIR. The high beads mucoadhesiveness was evidenced by in vitro and ex vivo tests. The erosion of beads was greater in acid media while swelling was more pronounced in pH 7.4. Drug release was dependent on pH in which samples 11H1-3, 11H1-5 and 41H1-3 released only 34%, 20% and 22% of ketoprofen in pH 1.2, while in pH 7.4 the drug release was sustained up to 360 min. Korsmeyer-Peppas model demonstrated that drug release occurred according to super case-II transport.
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Supelco
Ketoprofen, VETRANAL®, analytical standard
USP
Ketoprofen, United States Pharmacopeia (USP) Reference Standard
Ketoprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketoprofen, Pharmaceutical Secondary Standard; Certified Reference Material