Merck
  • Comparative method validation for closantel determination in cattle and sheep milk according to European Union Volume 8 and Veterinary International Conference on Harmonization guidelines.
Comparative method validation for closantel determination in cattle and sheep milk according to European Union Volume 8 and Veterinary International Conference on Harmonization guidelines.
Journal of chromatography. A (2014-03-25)
Mathias Devreese, An Maes, Siegrid De Baere, Patrick De Backer, Siska Croubels
ABSTRACT
A specific and sensitive LC-MS/MS method was developed for quantitative determination of closantel in bovine and ovine colostrum and tank milk. Sample preparation consisted of extracting milk samples with acetonitrile/acetone (80/20, v/v) followed by SPE clean-up with Oasis mixed anion exchange columns. After elution with 5% formic acid in acetonitrile and evaporation to dryness, the residue was reconstituted in acetonitrile and water. HPLC separation was achieved on a Zorbax Eclipse Plus C18 column and a gradient elution program with 1mM ammonium acetate in water and acetonitrile. For closantel determination in bovine milk, the method was validated according to EU Volume 8 guidelines whereas for ovine milk both EU Volume 8 and VICH GL49 criteria were applied. The linear range of the method is between 10 and 2000 μg/kg, the limit of quantification 10 μg/kg and limit of detection is 0.63 and 0.32 μg/kg for sheep colostrum and tank milk and 1.27 and 1.24 μg/kg for cattle colostrum and tank milk, respectively. Both guidelines cover a similar set of parameters (linearity, accuracy, precision, limit of detection and limit of quantification), although the acceptance criteria might differ (accuracy and precision) or no specific acceptability ranges are specified in neither guidelines (LOD and LOQ). For some parameters, only one of the guidelines indicates acceptance criteria: EU Volume 8 for applicability, practicability and susceptibility and VICH GL 49 for linearity, specificity and analyte stability.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Supelco
Acetone, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Acetone, natural, ≥97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Supelco
Tetrahydrofuran, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
Estimated to ship on 23 March 2025
MXP 1,249.00
1 kg
Estimated to ship on 23 March 2025
MXP 1,885.00
10 kg
Estimated to ship on 23 March 2025
MXP 8,138.00
20 kg
Estimated to ship on 23 March 2025
MXP 11,924.00
To order products, please contact your local dealer.