Merck
  • Development and validation of an HPTLC method for apigenin 7-O-glucoside in chamomile flowers and its application for fingerprint discrimination of chamomile-like materials.
Development and validation of an HPTLC method for apigenin 7-O-glucoside in chamomile flowers and its application for fingerprint discrimination of chamomile-like materials.
Journal of pharmaceutical and biomedical analysis (2015-01-13)
Etil Guzelmeric, Irena Vovk, Erdem Yesilada
ABSTRACT
Brewed tea of chamomile flowers (Matricaria recutita L.) (Asteraceae) has been extensively consumed for centuries due to either its pleasant taste or medicinal purposes. On the other hand, the major problem is difficulty in distinguishing the genuine specimen when supplying chamomile through nature-picking. Consequently flowers of other Asteraceae members resembling to chamomile in appearance may frequently be practiced by lay people or marketed in spice shops or bazaars. Evidently detection of such adulterations plays a vital role in terms of public health to avoid risk of toxicity (i.e. pyrazolidin alkaloids) and ineffective treatments (lack or insufficient concentration of the active constituents). This work presents either development and validation of a high performance thin-layer chromatographic (HPTLC) method for apigenin 7-O-glucoside which is one of the active markers in chamomile flowers or its application for the fingerprint discrimination of chamomile-like materials i.e. Anthemis spp., Bellis spp., Chrysanthemum sp. and Tanacetum sp. gathered by local people assuming as chamomile. Separation was performed on the silica gel 60 NH2 F254s HPTLC plates using the developing solvent system of ethyl acetate-formic acid-acetic acid-water (30:1.5:1.5:3, v/v/v/v). The proposed HPTLC method may also be a leading guide for the quality assessment of chamomile tea products on the market.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Acetic acid, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Acetone, analytical standard
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%