Merck
  • Development and validation of an ultra high performance liquid chromatography tandem mass spectrometry method for simultaneous determination of sulfonamides, quinolones and benzimidazoles in bovine milk.
Development and validation of an ultra high performance liquid chromatography tandem mass spectrometry method for simultaneous determination of sulfonamides, quinolones and benzimidazoles in bovine milk.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-06-03)
Xiao-Lin Hou, Guo Chen, Li Zhu, Ting Yang, Jian Zhao, Lei Wang, Yin-Liang Wu
ABSTRACT
A simple, sensitive and reliable analytical method was developed for the simultaneous determination of 38 veterinary drugs (18 sulfonamides, 11 quinolones and 9 benzimidazoles) and 8 metabolites of benzimidazoles in bovine milk by ultra high performance liquid chromatography-positive electrospray ionization tandem mass spectrometry (UHPLC-ESI-MS/MS). Samples were extracted with acidified acetonitrile, cleaned up with Oasis(®) MCX cartridges, and analyzed by LC-MS/MS on an Acquity UPLC(®) BEH C18 column with gradient elution. The method allows such multi-analyte measurements within a 13min runtime while the specificity is ensured through the MRM acquisition mode. The method was validated according to the European Commission Decision 2002/657/EC determining specificity, decision limit (CCα), detection capability (CCβ), recovery, precision, linearity and stability. For compounds which have MRLs in bovine milk, the CCα values fall into a range from 11 to 115μg/kg, and the CCβ values fall within a range of 12-125μg/kg. For compounds which have not MRLs in bovine milk, the CCα values fall into a range from 0.01 to 0.08μg/kg, and the CCβ values fall within a range of 0.02-0.11μg/kg. The mean recoveries of the 46 analytes were between 87 and 119%. The calculated RSD values of repeatability and within-laboratory reproducibility experiments were below 11% and 15% for the 46 compounds, respectively. The method was demonstrated to be suitable for the simultaneous determination of sulfonamides, quinolones and benzimidazoles in bovine milk.
Product Description
Supelco
Norfloxacin-d5, VETRANAL®, analytical standard
Supelco
Albendazole sulfone, VETRANAL®, analytical standard
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Mebendazole, VETRANAL®, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Mebendazole, European Pharmacopoeia (EP) Reference Standard
Fenbendazole, European Pharmacopoeia (EP) Reference Standard
USP
Mebendazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium acetate, for inorganic trace analysis, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Fenbendazole, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Fenbendazole, VETRANAL®, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Mebendazole, analytical standard, ≥98% (HPLC)
USP
Fenbendazole, United States Pharmacopeia (USP) Reference Standard