Merck
  • Development and validation of a hydrophilic interaction liquid chromatography with tandem mass spectrometry method for the simultaneous detection and quantification of etilefrine and oxilofrine in equine blood plasma and urine.
Development and validation of a hydrophilic interaction liquid chromatography with tandem mass spectrometry method for the simultaneous detection and quantification of etilefrine and oxilofrine in equine blood plasma and urine.
Journal of separation science (2014-08-22)
Noel Kong, Rong Yi, Sarah Zhao, Jasmeet Sandhu, Geoffrey Lam, Devan Loganathan, Barbara Morrissey
ABSTRACT
A sensitive hydrophilic interaction liquid chromatography coupled with tandem mass spectrometry method was developed and validated for the simultaneous detection and quantification of etilefrine and oxilofrine in equine blood plasma and urine. The method is highly sensitive and specific with good precision and accuracy. In plasma the limit of detection and limit of quantification are 0.03 and 0.1 ng/mL, respectively, for both analytes. In urine the limit of detection and limit of quantification are 0.3 and 1 ng/mL, respectively, for both analytes. The suitability of the method for doping control analysis in equine species is demonstrated by analyzing postadministration samples collected after a single intravenous administration of 50 mg etilefrine to a standardbred mare. Etilefrine was detected up to 120 h in urine and up to 48 h in plasma. Etilefrine is highly conjugated in equine urine whereas it exists in the free form in equine plasma. Therefore, enzyme hydrolysis prior to sample preparation is recommended for the detection and quantification of etilefrine and oxilofrine in equine urine.
Sigma-Aldrich
(1S,2R)-(+)-Ephedrine hydrochloride, 99%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
DL-Ephedrine hydrochloride, 99%
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Ephedrine hydrochloride (racemic), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)