Merck
  • Circulating vitamin D levels are associated with the presence and severity of coronary artery disease but not peripheral arterial disease in patients undergoing coronary angiography.
Circulating vitamin D levels are associated with the presence and severity of coronary artery disease but not peripheral arterial disease in patients undergoing coronary angiography.
Nutrition, metabolism, and cardiovascular diseases : NMCD (2015-02-03)
J Y Liew, S R Sasha, P J Ngu, J L Warren, J Wark, A M Dart, J A Shaw
ABSTRACT
To investigate the association between vitamin D levels, angiographic severity of coronary artery disease, arterial stiffness and degree of peripheral arterial disease (PAD) as assessed by ankle brachial index (ABI). 375 patients undergoing coronary angiography from November 2012 to September 2013 were recruited. Serum 25-hydroxyvitamin D (25OHD) levels were measured as were ABI and pulse wave velocity (PWV). Based on the findings of the coronary angiogram, patients were divided into subgroups: Absent, Single, Double and Triple Vessel Disease (as defined by >50% stenosis in each major coronary artery) 0.275 patients not taking vitamin D supplements were included in the analysis. Mean age was 66.0 ± 11.2 (mean ± SD) years. Levels of 25(OH)D were significantly lower in patients with CAD when compared to patients without CAD (57.0 ± 1.73 versus 70.1 ± 2.46 nmol/L; p < 0.01). One way ANOVA revealed triple vessel disease patients had significantly lower 25(OH)D levels when compared to single vessel disease patients (50.6 ± 2.84 nmol/L versus 61.3 ± 3.16 p < 0.01) and trended to be lower when compared to double vessel disease patients (50.6 ± 2.84 versus 59.0 ± 2.99 nmol/L; p = 0.07). Stepwise regression revealed that age, gender (male), hypertension, hyperlipidemia and 25(OH)D were significant predictors of CAD (p < 0.05). Vitamin D was the most significant predictor for CAD (p < 0.001) There was no correlation between 25(OH)D levels, ABI and PWV. Among patients presenting for coronary angiography, low serum 25-hydroxyvitamin D levels are associated with the presence and extent of angiographic CAD but not arterial stiffness or PAD.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%