Merck
  • Synthesis, chemical identification, antioxidant capacities and immunological evaluation studies of a novel silver(I) carbocysteine complex.
Synthesis, chemical identification, antioxidant capacities and immunological evaluation studies of a novel silver(I) carbocysteine complex.
Chemico-biological interactions (2014-07-08)
Samy M El-Megharbel, Reham Z Hamza, Moamen S Refat
ABSTRACT
A new silver-carbocysteine (Ccy-Ag) complex [Ag2(Ccy)2(H2O)2] has been synthesized and characterized by using a combination of FTIR, Raman, molar conductivity, (1)H NMR, electronic spectra, thermal analyses, X-ray powder diffraction (XRD) and scanning electron microscopy (SEM). The infrared spectrum of Ccy-Ag complex in comparison with carbocysteine ligand prove that Ccy behaves as monobasic bidentate chelate to the silver metal ions via the deprotonated carboxylate O atom. The assessments of Ccy and its complexation with Ag(+) in treating COPD, evaluating immune activities through measuring IL-8, TGF-β1, VEGF and TNF-α, antioxidant activities of (Ccy-Ag) complex by measuring SOD, MDA and GPX and bronchial asthma were discussed.
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, FT-IR grade, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Nitrogen, ≥99.998%