Skip to Content
Merck
All Photos(1)

Documents

449962

Sigma-Aldrich

Potassium bromide

anhydrous, powder, 99.999% trace metals basis

Synonym(s):

Bromide salt of potassium, Potassium monobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050405
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)

Assay

99.999% trace metals basis

form

powder

impurities

≤15.0 ppm Trace Metal Analysis

mp

734 °C (lit.)

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium bromide is an ionic salt that is used as a capping agent/medium in the preparation of nanomaterials such as ZrB2 and palladium nanoparticles.. It is also widely used as a source of bromine in various organic transformations.

Application

May be used for removal of peripheral membrane proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Microwave-Assisted Facile Synthesis of Palladium Nanoparticles in HEPES Solution and Their Size-Dependent Catalytic Activities to Suzuki Reaction
Wei Zhang, et al.
Journal of Nanoscience and Nanotechnology, 11, 7794-7801 (2011)
Preparation of ZrB2 by Reacting ZrCl4 with NaBH4 in Molten Potassium Bromide
SE Kravchenko, et al.
Inorganic Materials, 55, 458-461 (2019)
Igor L Dalinger et al.
Chemistry, an Asian journal, 10(9), 1987-1996 (2015-06-23)
A new family of energetic compounds, nitropyrazoles bearing a trinitromethyl moiety at the nitrogen atom of the heterocycle, was designed. The desirable high-energy dense oxidizers 3,4-dinitro- and 3,5-dinitro-1-(trinitromethyl)pyrazoles were synthesized in good yields by destructive nitration of the corresponding 1-acetonylpyrazoles.
Pascal Miéville et al.
Magnetic resonance in chemistry : MRC, 49(11), 689-692 (2011-10-18)
As previously demonstrated by Thurber and Tycko, the peak position of (79)Br in potassium bromide (KBr) allows one to determine the temperature of a spinning sample. We propose to adapt the original design by using a compact KBr tablet placed
Hao-Long Zhou et al.
Nature communications, 6, 6917-6917 (2015-04-22)
The dynamic behaviours of host frameworks and guest molecules have received much attention for their great relevance with smart materials, but little has been developed to control or understand the host-guest interplay. Here we show that the confined guest can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service