Merck
  • A 3D-printed device for a smartphone-based chemiluminescence biosensor for lactate in oral fluid and sweat.
A 3D-printed device for a smartphone-based chemiluminescence biosensor for lactate in oral fluid and sweat.
The Analyst (2014-10-25)
Aldo Roda, Massimo Guardigli, Donato Calabria, Maria Maddalena Calabretta, Luca Cevenini, Elisa Michelini
ABSTRACT
Increasingly, smartphones are used as portable personal computers, revolutionizing communication styles and entire lifestyles. Using 3D-printing technology we have made a disposable minicartridge that can be easily prototyped to turn any kind of smartphone or tablet into a portable luminometer to detect chemiluminescence derived from enzyme-coupled reactions. As proof-of-principle, lactate oxidase was coupled with horseradish peroxidase for lactate determination in oral fluid and sweat. Lactate can be quantified in less than five minutes with detection limits of 0.5 mmol L(-1) (corresponding to 4.5 mg dL(-1)) and 0.1 mmol L(-1) (corresponding to 0.9 mg dL(-1)) in oral fluid and sweat, respectively. A smartphone-based device shows adequate analytical performance to offer a cost-effective alternative for non-invasive lactate measurement. It could be used to evaluate lactate variation in relation to the anaerobic threshold in endurance sport and for monitoring lactic acidosis in critical-care patients.
Product Description
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioReagent, suitable for electrophoresis, 98.0-102.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SAFC
L-Histidine monohydrochloride monohydrate
Sigma-Aldrich
L-Histidine monohydrochloride monohydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Histidine monohydrochloride monohydrate, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Histidine hydrochloride monohydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, puriss. p.a., ACS reagent, ≥99.0% (T)