Skip to Content
Merck
All Photos(1)

Key Documents

08582

Millipore

Urea solution

suitable for microbiology, 40% in H2O

Synonym(s):

Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
MDL number:
UNSPSC Code:
41171614
PubChem Substance ID:
NACRES:
NA.85

Agency

according to ISO 6579-1:2017

form

liquid

shelf life

limited shelf life, expiry date on the label

concentration

40% in H2O

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Proteus spp.

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Filter sterilized urea solution recommended for detection of urease activity.

Application

for detection and enumeration of coliform organisms from chlorinated water by membrane filter technique

Components

(per vial, sufficient for 100 mL medium):
Urea 2.0 g, Distilled water 5.0 ml

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G L French et al.
Lancet (London, England), 2(8551), 117-119 (1987-07-18)
Tuberculostearic acid, a structural component of Mycobacterium tuberculosis, was identified by gas chromatography/mass spectrometry with selected ion monitoring in cerebrospinal fluid (CSF) from 13 patients with proven and 8 out of 9 patients with suspected tuberculous meningitis; the negative result
A mechanically and electrically self-healing supercapacitor.
Hua Wang et al.
Advanced materials (Deerfield Beach, Fla.), 26(22), 3638-3643 (2014-02-21)
Oliver C Witard et al.
Brain, behavior, and immunity, 39, 211-219 (2013-10-15)
The present study examined whether a high protein diet prevents the impaired leukocyte redistribution in response to acute exercise caused by a large volume of high-intensity exercise training. Eight cyclists (VO2max: 64.2±6.5mLkg(-1)min(-1)) undertook two separate weeks of high-intensity training while
Sreenivasa C Ramaiahgari et al.
Archives of toxicology, 88(5), 1083-1095 (2014-03-07)
Immortalized hepatocyte cell lines show only a weak resemblance to primary hepatocytes in terms of gene expression and function, limiting their value in predicting drug-induced liver injury (DILI). Furthermore, primary hepatocytes cultured on two-dimensional tissue culture plastic surfaces rapidly dedifferentiate
Guillaume Carissimo et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(2), E176-E185 (2014-12-31)
Arboviruses are transmitted by mosquitoes and other arthropods to humans and animals. The risk associated with these viruses is increasing worldwide, including new emergence in Europe and the Americas. Anopheline mosquitoes are vectors of human malaria but are believed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service