Merck
  • Interfacial assembly of dendritic microcapsules with host-guest chemistry.
Interfacial assembly of dendritic microcapsules with host-guest chemistry.
Nature communications (2014-12-17)
Yu Zheng, Ziyi Yu, Richard M Parker, Yuchao Wu, Chris Abell, Oren A Scherman
ABSTRACT
The self-assembly of nanoscale materials to form hierarchically ordered structures promises new opportunities in drug delivery, as well as magnetic materials and devices. Herein, we report a simple means to promote the self-assembly of two polymers with functional groups at a water-chloroform interface using microfluidic technology. Two polymeric layers can be assembled and disassembled at the droplet interface using the efficiency of cucurbit[8]uril (CB[8]) host-guest supramolecular chemistry. The microcapsules produced are extremely monodisperse in size and can encapsulate target molecules in a robust, well-defined manner. In addition, we exploit a dendritic copolymer architecture to trap a small hydrophilic molecule in the microcapsule skin as cargo. This demonstrates not only the ability to encapsulate small molecules but also the ability to orthogonally store both hydrophilic and hydrophobic cargos within a single microcapsule. The interfacially assembled supramolecular microcapsules can benefit from the diversity of polymeric materials, allowing for fine control over the microcapsule properties.
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
2-Hydroxyethyl methacrylate, embedding medium (for microscopy)
Sigma-Aldrich
2-Cyano-2-propyl benzodithioate, >97% (HPLC)
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Methyl methacrylate, European Pharmacopoeia (EP) Reference Standard
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
2-(Dimethylamino)ethyl methacrylate, contains 700-1000 ppm monomethyl ether hydroquinone as inhibitor, 98%
Supelco
Hexane, analytical standard
Sigma-Aldrich
Rhodamine B isothiocyanate, mixed isomers
Sigma-Aldrich
Fluorescein o-acrylate, 95%
Sigma-Aldrich
2-Naphthyl isocyanate, 97%