Merck
  • Disturbed B cell homeostasis in newly diagnosed giant cell arteritis and polymyalgia rheumatica.
Disturbed B cell homeostasis in newly diagnosed giant cell arteritis and polymyalgia rheumatica.
Arthritis & rheumatology (Hoboken, N.J.) (2014-03-14)
Kornelis S M van der Geest, Wayel H Abdulahad, Paulina Chalan, Abraham Rutgers, Gerda Horst, Minke G Huitema, Mirjam P Roffel, Caroline Roozendaal, Philip M Kluin, Nicolaas A Bos, Annemieke M H Boots, Elisabeth Brouwer
ABSTRACT
Several lines of evidence indicate that B cells may be involved in the immunopathology of giant cell arteritis (GCA) and polymyalgia rheumatica (PMR). This study was undertaken to examine the distribution of defined B cell subsets, including effector B (Beff) cells and regulatory B (Breg) cells, in patients with GCA and patients with PMR before and after corticosteroid treatment. Circulating B cells were analyzed in 34 newly diagnosed, untreated patients with GCA or PMR, and in 44 followup samples from patients with GCA or PMR who received corticosteroids for 2 weeks or 3 months. For comparison, 40 age-matched healthy controls and 11 rheumatoid arthritis (RA) patients were included. Serum BAFF levels were determined, and temporal arteries were studied by immunohistochemistry. Patients newly diagnosed as having GCA or PMR, but not patients with RA, had decreased numbers of circulating B cells compared to healthy controls. B cell numbers recovered rapidly in treated patients with GCA and PMR in remission. This recovery was not achieved by compensatory hyperproliferation or enhanced bone marrow production. B cell numbers inversely correlated with erythrocyte sedimentation rates, C-reactive protein levels, and serum BAFF levels. Tumor necrosis factor α-positive Beff cells, but not interleukin-10 (IL-10)-positive Breg cells, were decreased in patients newly diagnosed as having GCA or PMR. Following treatment, circulating numbers of Beff cells normalized. The returning Beff cells demonstrated an enhanced capacity to produce IL-6. Few B cells were found in temporal artery biopsy specimens from GCA patients. We show for the first time that the distribution of B cells is highly disturbed in GCA and PMR and that B cells likely contribute to the enhanced IL-6 response in both diseases.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Sigma-Aldrich
BAFF active human, Animal-component free, recombinant, expressed in Nicotiana, >97% (SDS-PAGE)
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
BAFF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Supelco
Ammonium ion solution for ISE, for ion-selective electrodes, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)