Merck
  • Synthesis of novel phosphonic-type activity-based probes for neutrophil serine proteases and their application in spleen lysates of different organisms.
Synthesis of novel phosphonic-type activity-based probes for neutrophil serine proteases and their application in spleen lysates of different organisms.
Chembiochem : a European journal of chemical biology (2014-09-23)
Renata Grzywa, Ewa Burchacka, Maria Łęcka, Łukasz Winiarski, Maciej Walczak, Agnieszka Łupicka-Słowik, Magdalena Wysocka, Timo Burster, Kamila Bobrek, Keri Csencsits-Smith, Adam Lesner, Marcin Sieńczyk
ABSTRACT
Neutrophils are a type of granulocyte important in the "first line of defense" of the innate immune system. Upon activation, they facilitate the destruction of invading microorganisms by the production of superoxide radicals, as well as the release of the enzymatic contents of their lysozymes. These enzymes include specific serine proteases: cathepsin G, neutrophil elastase, proteinase 3, as well as the recently discovered neutrophil serine protease 4 (NSP4). Under normal conditions, the proteolytic activity of neutrophil proteases is tightly regulated by endogenous serpins; however, this mechanism can be subverted during tissue stress, thereby resulting in the uncontrolled activity of serine proteases, which induce chronic inflammation and subsequent pathology. Herein, we describe the development of low-molecular-weight activity-based probes that specifically target the active sites of neutrophil proteases.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Amylamine, ≥99%, FG
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Amylamine, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Amylamines, mixture of isomers, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Millipore
Bifido Selective Supplement B, suitable for microbiology