Merck
  • Timing matters: sensitivity of Daphnia magna dormant eggs to fenoxycarb exposure depends on embryonic developmental stage.
Timing matters: sensitivity of Daphnia magna dormant eggs to fenoxycarb exposure depends on embryonic developmental stage.
Aquatic toxicology (Amsterdam, Netherlands) (2014-12-30)
Sabine Navis, Aline Waterkeyn, Adinda Putman, Luc De Meester, Guido Vanermen, Luc Brendonck
ABSTRACT
Although Daphnia magna is a key species in many lentic freshwater ecosystems and is commonly used as model organism in ecology and ecotoxicology, very little is known about the effects of chemicals on their dormant life stages. Dormant eggs (ephippia) are produced when environmental conditions deteriorate, and Daphnia switch from clonal to sexual reproduction. Ephippia produced over different growing seasons can accumulate in the sediment of ponds and lakes, where they can be exposed to pesticides and other (anthropogenic) stressors. In the present study, we have investigated the effects of pesticide exposure on dormant eggs at different embryonic developmental stages and evaluated the degree of protection against pollution provided by the ephippial case. We therefore conducted a hatching experiment in which decapsulated and encapsulated dormant eggs were exposed to an insect growth regulator (fenoxycarb) at different stages during their development, both before and after activation of the eggs. In addition, we developed an analytical method to measure fenoxycarb concentrations in the dormant eggs. Fenoxycarb negatively affected development and hatching success and changed the timing of hatching in activated and in dormant eggs. Hatching characteristics as well as fenoxycarb concentrations inside the eggs differed significantly between exposure treatments. Final stages of embryonic development were most sensitive to pesticide exposure and had the highest tissue concentrations of fenoxycarb. Tissue concentrations did not differ significantly between decapsulated and encapsulated eggs, suggesting that the ephippial case offers limited or no direct protection against pesticide exposure. With this study we provide new evidence showing that pesticides can bioconcentrate in and affect D. magna dormant eggs. The severity of the effects on developing embryos depends on the timing of pesticide exposure. Our results stress the importance of considering the full life-cycle of model organisms used in ecotoxicological studies, since these are ultimately aimed at assessing risks of chemical exposure on natural aquatic ecosystems.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%