Merck
  • TLR4 inhibition impairs bacterial clearance in a therapeutic setting in murine abdominal sepsis.
TLR4 inhibition impairs bacterial clearance in a therapeutic setting in murine abdominal sepsis.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2014-08-15)
Miriam H P van Lieshout, Tom van der Poll, Cornelis van't Veer
ABSTRACT
To investigate the therapeutic effect of E5564 (a clinically used TLR4 inhibitor) in murine abdominal sepsis elicited by intraperitoneal infection with a highly virulent Escherichia coli in the context of concurrent antibiotic therapy. Mice were infected with different doses (~2 × 10(4)-2 × 10(6) CFU) of E. coli O18:K1 and treated after 8 h with ceftriaxone 20 mg/kg i.p. combined with either E5564 10 mg/kg i.v. or vehicle. For survival studies this treatment was repeated every 12 h. Bacterial loads and inflammatory parameters were determined after 20 h in peritoneal lavage fluid, blood, liver and lung tissue. Plasma creatinin, AST, ALT and LDH were determined to assess organ injury. E5564 impaired bacterial clearance under the antibiotic regime after infection with a low dose E. coli (1.7 × 10(4) CFU) while renal function was slightly preserved. No differences were observed in bacterial load and organ damage after infection with a tenfold higher (1.7 × 10(5) E. coli) bacterial dose. While treatment with E5564 slightly attenuated inflammatory markers provoked by the sublethal doses of 104-105 E. coli under the antibiotic regime, it did not affect lethality evoked by infection with 1.7 × 106 E. coli. The impact of TLR4 inhibition during abdominal sepsis by virulent E. coli bacteria is only beneficial at low infection grade at cost of bactericidal activity.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mouse E-Selectin ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human E-Selectin ELISA Kit, for serum, plasma, cell culture supernatants and urine