Merck
  • Depression and synaptic zinc regulation in Alzheimer disease, dementia with lewy bodies, and Parkinson disease dementia.
Depression and synaptic zinc regulation in Alzheimer disease, dementia with lewy bodies, and Parkinson disease dementia.
The American journal of geriatric psychiatry : official journal of the American Association for Geriatric Psychiatry (2014-06-24)
David R Whitfield, Julie Vallortigara, Amani Alghamdi, Tibor Hortobágyi, Clive Ballard, Alan J Thomas, John T O'Brien, Dag Aarsland, Paul T Francis
ABSTRACT
Depression is a common symptom in dementia with Lewy bodies (DLB), Parkinson disease dementia (PDD), and Alzheimer disease (AD), yet its molecular basis remains unclear and current antidepressants do not appear to be effective. Cerebral zinc has been implicated in depression and synaptic dysfunction. We investigated the relationship between synaptic zinc regulation (for which zinc transporter 3 [ZnT3] is responsible) and depression in a large clinicopathologic study. We examined brains from people with PDD (N = 29), DLB (N = 27), and AD (N = 15) and comparison subjects without depression or dementia (N = 24). Individuals were categorized according to the presence and severity of depression (on a scale of 0-3) based on standardized assessments during life (principally Neuropsychiatric Inventory). Western blotting was used to determine ZnT3 levels in Brodmann area 9 (BA9), and regression analysis was used to determine the relationship between ZnT3 and depression. Reductions in ZnT3 in BA9 were significantly associated with elevated depression scores in the study cohort (β = -0.351, df = 93, t = -3.318 p = 0.0004). This association remained when only individuals with DLB, PDD, and no dementia or depression were examined (β = -0.347, df = 78, t = -3.271, p = 0.002) or only individuals with AD and no dementia or depression were examined (β = -0.433, df = 37, t = -2.924, p = 0.006). Although decreased zinc levels have been implicated in the genesis of depression in animal models and in major depressive disorder in humans, this study provides the first evidence of a role for zinc in depression in people with dementia and highlights zinc metabolism as a therapeutic target.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder