Merck
  • Cyclocoagulation of the ciliary bodies by high-intensity focused ultrasound: a 12-month multicenter study.
Cyclocoagulation of the ciliary bodies by high-intensity focused ultrasound: a 12-month multicenter study.
Investigative ophthalmology & visual science (2015-01-22)
Philippe Denis, Florent Aptel, Jean-François Rouland, Jean-Philippe Nordmann, Yves Lachkar, Jean-Paul Renard, Eric Sellem, Christophe Baudouin, Alain Bron
ABSTRACT
To evaluate the safety and efficacy of high-intensity focused ultrasound (HIFU) cyclocoagulation in reducing intraocular pressure (IOP) in patients with refractory glaucoma by using a novel miniaturized delivery device (EyeOP1). We conducted a 12-month open-label multicenter prospective study (EyeMUST1 Study). Patients with primary (primary open-angle glaucoma [POAG]) or secondary refractory glaucoma were treated in two groups depending on the duration of each ultrasound shot (group 1: 4 seconds; group 2: 6 seconds). The primary efficacy outcome was based on IOP reduction at 6 and 12 months. Fifty-two patients were enrolled: 36 (69%) had POAG and 16 (31%) had secondary glaucoma. Group 1 (n = 24) and group 2 (n = 28) had similar demographics and baseline characteristics. In group 1, IOP was reduced from a mean preoperative value of 29.7 ± 7.7 mm Hg (n = 3.5 glaucoma medications) to a mean postoperative value of 21.3 ± 6.7 mm Hg (n = 3.5 glaucoma medications) and 20.1 ± 6.7 mm Hg (n = 3.2 glaucoma medications) at 6 and 12 months, respectively. In group 2, IOP was reduced from a mean preoperative value of 29.0 ± 7.4 mm Hg (n = 3.3 glaucoma medications) to a mean postoperative value of 20.2 ± 7.4 mm Hg (n = 3.4 glaucoma medications) and 18.5 ± 6.6 mm Hg (n = 3.5 glaucoma medications) at 6 and 12 months, respectively. At 12 months, the IOP reduction was sustained in both groups (32% IOP reduction in group 1 and 36% IOP reduction in group 2). The overall tolerance of the technique was good, with no serious adverse events. The new miniaturized HIFU EyeOP1 delivery device seems to be effective in decreasing IOP in patients with refractory glaucoma. The technology offers a good safety profile. (ClinicalTrials.gov number, NCT01338467.).
Sigma-Aldrich
Tobramycin, Aminoglycoside antibiotic
Tobramycin, European Pharmacopoeia (EP) Reference Standard
USP
Tobramycin, United States Pharmacopeia (USP) Reference Standard
Supelco
Tobramycin, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Flurbiprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Flurbiprofen, cyclooxygenase inhibitor
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Flurbiprofen, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard