Skip to Content
Merck
All Photos(1)

Key Documents

46165

Supelco

Dexamethasone

VETRANAL®, analytical standard

Synonym(s):

(11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
Beilstein:
2066651
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

262-264 °C (lit.)

application(s)

clinical

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

InChI

1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1

InChI key

UREBDLICKHMUKA-CXSFZGCWSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Dexamethasone is a synthetic glucocorticoid and cortisol analogue. It is used in the suppression of adrenocorticotropic hormone (ACTH) and cortisol production in normal subjects.[1]

Application

Dexamethasone may be used as a reference standard in the determination of dexamethasone in biological samples using high-performance liquid chromatography (HPLC).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Glucocorticoid anti-inflammatory agent
Glucocorticoid anti-inflammatory agent. Regulates T cell survival, growth, and differentiation. Inhibits the induction of nitric oxide synthase.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved HPLC method for simultaneous analysis of cortisol, 11-deoxycortisol, prednisolone, methylprednisolone, and dexamethasone in serum and urine.
McWhinney CB, et al.
Clinical Chemistry, 42(6), 979-981 (1996)
Tietz Textbook of Clinical Chemistry and Molecular Diagnostics (2012)
Erich Piovan et al.
Cancer cell, 24(6), 766-776 (2013-12-03)
Glucocorticoid resistance is a major driver of therapeutic failure in T cell acute lymphoblastic leukemia (T-ALL). Here, we identify the AKT1 kinase as a major negative regulator of the NR3C1 glucocorticoid receptor protein activity driving glucocorticoid resistance in T-ALL. Mechanistically
Yukiko Kitase et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 25(12), 2657-2668 (2010-06-26)
Glucocorticoids are known to induce osteocyte apoptosis, whereas mechanical loading has been shown to sustain osteocyte viability. Here we show that mechanical loading in the form of fluid-flow shear stress blocks dexamethasone-induced apoptosis of osteocyte-like cells (MLO-Y4). Prostaglandin E(2) (PGE(2)
Lotfi Benboubker et al.
The New England journal of medicine, 371(10), 906-917 (2014-09-04)
The combination melphalan-prednisone-thalidomide (MPT) is considered a standard therapy for patients with myeloma who are ineligible for stem-cell transplantation. However, emerging data on the use of lenalidomide and low-dose dexamethasone warrant a prospective comparison of the two approaches. We randomly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service