Merck
  • Hind limb unloading, a model of spaceflight conditions, leads to decreased B lymphopoiesis similar to aging.
Hind limb unloading, a model of spaceflight conditions, leads to decreased B lymphopoiesis similar to aging.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2014-11-08)
Chloé Lescale, Véronique Schenten, Dounia Djeghloul, Meriem Bennabi, Fanny Gaignier, Katleen Vandamme, Catherine Strazielle, Isabelle Kuzniak, Hervé Petite, Christine Dosquet, Jean-Pol Frippiat, Michele Goodhardt
ABSTRACT
Within the bone marrow, the endosteal niche plays a crucial role in B-cell differentiation. Because spaceflight is associated with osteoporosis, we investigated whether changes in bone microstructure induced by a ground-based model of spaceflight, hind limb unloading (HU), could affect B lymphopoiesis. To this end, we analyzed both bone parameters and the frequency of early hematopoietic precursors and cells of the B lineage after 3, 6, 13, and 21 d of HU. We found that limb disuse leads to a decrease in both bone microstructure and the frequency of B-cell progenitors in the bone marrow. Although multipotent hematopoietic progenitors were not affected by HU, a decrease in B lymphopoiesis was observed as of the common lymphoid progenitor (CLP) stage with a major block at the progenitor B (pro-B) to precursor B (pre-B) cell transition (5- to 10-fold decrease). The modifications in B lymphopoiesis were similar to those observed in aged mice and, as with aging, decreased B-cell generation in HU mice was associated with reduced expression of B-cell transcription factors, early B-cell factor (EBF) and Pax5, and an alteration in STAT5-mediated IL-7 signaling. These findings demonstrate that mechanical unloading of hind limbs results in a decrease in early B-cell differentiation resembling age-related modifications in B lymphopoiesis.
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Supelco
Corticosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Corticosterone, ≥92%
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Supelco
Ammonium ion solution for ISE, for ion-selective electrodes, 1000 mg/kg N, analytical standard (for ion-selective electrodes)