Merck
  • Genetic removal of matrix metalloproteinase 9 rescues the symptoms of fragile X syndrome in a mouse model.
Genetic removal of matrix metalloproteinase 9 rescues the symptoms of fragile X syndrome in a mouse model.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-07-25)
Harpreet Sidhu, Lorraine E Dansie, Peter W Hickmott, Douglas W Ethell, Iryna M Ethell
ABSTRACT
Fmr1 knock-out (ko) mice display key features of fragile X syndrome (FXS), including delayed dendritic spine maturation and FXS-associated behaviors, such as poor socialization, obsessive-compulsive behavior, and hyperactivity. Here we provide conclusive evidence that matrix metalloproteinase-9 (MMP-9) is necessary to the development of FXS-associated defects in Fmr1 ko mice. Genetic disruption of Mmp-9 rescued key aspects of Fmr1 deficiency, including dendritic spine abnormalities, abnormal mGluR5-dependent LTD, as well as aberrant behaviors in open field and social novelty tests. Remarkably, MMP-9 deficiency also corrected non-neural features of Fmr1 deficiency-specifically macroorchidism-indicating that MMP-9 dysregulation contributes to FXS-associated abnormalities outside the CNS. Further, MMP-9 deficiency suppressed elevations of Akt, mammalian target of rapamycin, and eukaryotic translation initiation factor 4E phosphorylation seen in Fmr1 ko mice, which are also associated with other autistic spectrum disorders. These findings establish that MMP-9 is critical to the mechanisms responsible for neural and non-neural aspects of the FXS phenotype.
Sigma-Aldrich
Gelatin-Agarose, saline suspension
Millipore
Protein A–Agarose, lyophilized powder
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Mouse TIMP-1 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat TIMP-1 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat TIMP-1 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human TIMP-1 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human TIMP-1 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse TIMP-1 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Sample Buffer, Laemmli 2× Concentrate
Sigma-Aldrich
4-Aminophenylmercuric acetate, ≥90% (titration)
Sigma-Aldrich
Anti-PSD-95 (Ab-2) Mouse mAb (7E3-1B8), liquid, clone 7E3-1B8, Calbiochem®
Sigma-Aldrich
Anti-Synapsin I Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-FAK Antibody, Upstate®, from rabbit