Merck
  • Expression of organic osmolyte transporters in cultured rat astrocytes and rat and human cerebral cortex.
Expression of organic osmolyte transporters in cultured rat astrocytes and rat and human cerebral cortex.
Archives of biochemistry and biophysics (2014-07-09)
Jessica Oenarto, Boris Görg, Michael Moos, Hans-Jürgen Bidmon, Dieter Häussinger
ABSTRACT
This study characterizes the expression of the osmolyte transporters betaine/γ-amino-n-butyric acid (GABA) transporter (BGT-1), the taurine transporter (TauT) and the sodium-dependent myo-inositol transporter (SMIT) in various rat brain cells in culture and in rat and human cerebral cortex in situ. Osmolyte transporter expression greatly differed between cultured brain cells with highest mRNA expression levels for SMIT in astrocytes and TauT in neurons. BGT-1 mRNA and protein were expressed in microglia but not in astrocytes and neurons. In rat and human cerebral cortex, SMIT was expressed in astrocytes and TauT was found in neurons. Osmolyte transporter expression was subject to regulation by factors relevant for hepatic encephalopathy (HE). Hypoosmolarity, NH4Cl (0.5-5 mmol/l), diazepam (10 μmol/l) and TNFα (10 ng/ml) time-dependently decreased mRNA expression of SMIT and/or TauT in cultured astrocytes. NH4Cl-induced SMIT/TauT mRNA expression changes were sensitive to inhibitors of glutamine synthetase and NADPH oxidase. In rat cerebral cortex, SMIT mRNA expression decreased after portal vein ligation or ammonium acetate injection probably due to astrocyte swelling in these HE animal models. It is concluded that osmolyte transporters are heterogeneously expressed in brain and are subject to regulation by HE-relevant factors.
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Supelco
L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
SAFC
L-Methionine
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Diazepam
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®