Merck
Two-dimensional nanoarchitectures for lithium storage.
Advanced materials (Deerfield Beach, Fla.) (2012-04-17)
Jiehua Liu, Xue-Wei Liu
ABSTRACT
Two dimensional nanoarchitectures are of great interest in lithium storage for energy-storage devices, in particular lithium-ion batteries, due to its shortened paths for fast lithium ion diffusion and large exposed surface offering more lithium-insertion channels. Their competitive lithium-storage features provide huge potentials to develop next-generation high-performance lithium-ion batteries. This review is devoted to the recent progress in the fabrication of innovative 2D structures with various synthetic strategies and their applications for lithium storage in lithium-ion batteries. These 2D architectures are categorized into six styles, i.e., nanoporous nanosheets, ultrathin nanosheets, flower-like structures assembled by nanosheets, sandwich-like nanosheets, corrugated nanosheets, and nanosheets with specific facets. Based on the lithium-storage manner, we further summerized their electrochemical performance for lithium storage with four classified themes including surface Li storage, zero or low-strain Li storage, volume-variation Li storage and synergic-effect Li storage. Finally, the outlook and perspective on 2D lithium-storage materials is concisely provided.
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Lithium, foil, 25x100mm, thickness 0.6mm, as rolled, 99.9%
Lithium, foil, not light tested, 38x200mm, thickness 0.20mm, as rolled, 99.9%
Sigma-Aldrich
Lithium, foil, not light tested, 38x500mm, thickness 0.20mm, as rolled, 99.9%