Skip to Content
Merck
All Photos(2)

Documents

GF47159826

Lithium

foil, 25x100mm, thickness 0.6mm, as rolled, 99.9%

Synonym(s):

Lithium, LI000260

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li
CAS Number:
Molecular Weight:
6.94
EC Number:
MDL number:
UNSPSC Code:
12141803
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

foil

manufacturer/tradename

Goodfellow 471-598-26

resistivity

9.446 μΩ-cm, 20°C

size × thickness

25 x 100 mm × 0.6 mm

bp

1342 °C (lit.)

mp

180 °C (lit.)

density

0.534 g/mL at 25 °C (lit.)

SMILES string

[Li]

InChI

1S/Li

InChI key

WHXSMMKQMYFTQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium (Li) is a highly reactive alkali metal known for its lightweight and electrochemical properties. This product is available in foil form and has a resistivity of 9.446 μΩ-cm at 20°C. With an assay of 99.9%, lithium is characterized by its low melting point of 180 °C and a density of 0.534 g/mL at 25 °C. Lithium is extensively utilized in the production of rechargeable lithium-ion batteries, which power a wide range of electronic devices, including smartphones, laptops, and electric vehicles. Its lightweight nature and high energy density make it an ideal choice for energy storage applications. Additionally, lithium compounds are used in the manufacturing of ceramics, glass, and lubricating greases, enhancing their thermal and mechanical properties.

Application

  • A retrospective on lithium-ion batteries: This article reviews the contributions in the development of lithium-ion batteries, discussing inspirational insights to guide future breakthroughs in battery technology (J Xie, YC Lu, 2020).
  • Guidelines and trends for next-generation rechargeable lithium and lithium-ion batteries: This review discusses the limitations of commercial lithium-ion batteries and explores the directions for next-generation batteries (F Wu, J Maier, Y Yu, 2020).
  • Prospects for lithium-ion batteries and beyond—a 2030 vision: This article explores the future of lithium-ion batteries and discusses current strategies to improve these and next-generation battery technologies (CP Grey, DS Hall, 2020).
  • Lithium brines: A global perspective: This study offers a global perspective on lithium brines, discussing the critical and technologically important roles of lithium, which is in increasing demand (LA Munk, SA Hynek, DC Bradley, D Boutt, K Labay, 2016).

Legal Information

Product of Goodfellow

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingying Lu et al.
Nature materials, 13(10), 961-969 (2014-08-12)
Rechargeable lithium, sodium and aluminium metal-based batteries are among the most versatile platforms for high-energy, cost-effective electrochemical energy storage. Non-uniform metal deposition and dendrite formation on the negative electrode during repeated cycles of charge and discharge are major hurdles to
Susan W Lehmann et al.
Journal of affective disorders, 146(2), 151-157 (2012-09-19)
Lithium has been reported to induce hypercalcemia and hyperparathyroidism, yet few studies have examined the impact on older patients. We therefore undertook this review and report our findings. We undertook a systematic review of articles on lithium-associated hypercalcemia and/or hyperparathyroidism
Rebecca F McKnight et al.
Lancet (London, England), 379(9817), 721-728 (2012-01-24)
Lithium is a widely used and effective treatment for mood disorders. There has been concern about its safety but no adequate synthesis of the evidence for adverse effects. We aimed to undertake a clinically informative, systematic toxicity profile of lithium.
Xianglong Li et al.
Nanoscale, 5(19), 8864-8873 (2013-08-15)
The implementation of silicon (Si) in practical lithium ion battery electrodes has been hindered due to its large volume change and consequent structural and interfacial instabilities. Coating nanostructured Si with a second phase (e.g., carbon (C)) represents a very promising
Zhi Yang Tam et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 69(7), 810-820 (2014-01-09)
Aging has been associated with the accumulation of damages in molecules and organelles in cells, particularly mitochondria. The rate of damage accumulation is closely tied to the turnover of the affected cellular components. Perturbing mitochondrial turnover has been shown to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service