Merck
  • [beta-Cyclodextrin bonded stationary phases for chiral separation in HPLC].
[beta-Cyclodextrin bonded stationary phases for chiral separation in HPLC].
Se pu = Chinese journal of chromatography (2003-01-30)
H Chen, Q S Wang
ABSTRACT
A review of beta-cyclodextrin bonded stationary phases in chiral liquid chromatography is given, focusing on design and synthesis of beta-cyclodextrin bonded chiral stationary phases. Fifty-three references are presented in this paper.
Sigma-Aldrich
β-Cyclodextrin, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, ≥98.0% (HPLC)
Sigma-Aldrich
β-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥95.0% (HPLC)
Sigma-Aldrich
β-Cyclodextrin, ≥97%
Sigma-Aldrich
β-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
β-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany
USP
Beta Cyclodextrin, United States Pharmacopeia (USP) Reference Standard
Betadex, European Pharmacopoeia (EP) Reference Standard
Supelco
Beta-Cyclodextrin, Pharmaceutical Secondary Standard; Certified Reference Material