Merck
  • 11C-acetate PET/CT for metabolic characterization of multiple myeloma: a comparative study with 18F-FDG PET/CT.
11C-acetate PET/CT for metabolic characterization of multiple myeloma: a comparative study with 18F-FDG PET/CT.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2014-03-29)
Chi-lai Ho, Sirong Chen, Yim Lung Leung, Thomas Cheng, Ka-nin Wong, Shing Kee Cheung, Raymond Liang, Chor Sang Chim
ABSTRACT
We prospectively compared (11)C-acetate with (18)F-FDG in a PET/CT evaluation of multiple myeloma (MM), specifically on diagnostic accuracy, identification of high-risk patients, and monitoring of treatment response. Dual-tracer PET/CT was performed on 35 pathologically and clinically confirmed and untreated patients (26 with symptomatic MM, 5 with smoldering MM, and 4 with monoclonal gammopathy of unknown significance) and 20 individuals with normal marrow. (11)C-acetate showed significant incremental value over (18)F-FDG (84.6% vs. 57.7%) for positively identifying patients with diffuse and focal symptomatic MM, and was negative in patients with indolent smoldering MM and monoclonal gammopathy of unknown significance. Three functional parameters-number of (11)C-acetate-avid and (18)F-FDG-avid focal bone lesions and (11)C-acetate general marrow activity-strongly correlated with β-2-microglobulin as surrogate imaging markers of tumor burden. After induction chemotherapy, the metabolic change in (11)C-acetate general marrow activity correlated with clinical response. Metabolic characterization of MM in diagnosis, risk stratification, and treatment monitoring can be done more accurately by assessing lipid metabolism with (11)C-acetate than by assessing glucose metabolism with (18)F-FDG.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
2-Fluoro-2-deoxy-D-glucose, glycosylation inhibitor, glucose analog
Carbon, rod, 150mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 100mm, diameter 3.0mm, graphite, 100%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Millipore
Activated charcoal, suitable for GC
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Sigma-Aldrich
Activated charcoal, acid-washed with phosphoric and sulfuric acids
Carbon, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Carbon, rod, 50mm, diameter 0.5mm, graphite, 99.95%
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)