Merck
  • Valproic acid suppresses Nrf2/Keap1 dependent antioxidant protection through induction of endoplasmic reticulum stress and Keap1 promoter DNA demethylation in human lens epithelial cells.
Valproic acid suppresses Nrf2/Keap1 dependent antioxidant protection through induction of endoplasmic reticulum stress and Keap1 promoter DNA demethylation in human lens epithelial cells.
Experimental eye research (2014-02-15)
Periyasamy Palsamy, Keshore R Bidasee, Toshimichi Shinohara
ABSTRACT
Recent epidemiological studies confirm the prevalence of cataract in epileptic patients. Similarly, the drugs used to treat epilepsy also show the connection with increased cataract formation. In this present study, we investigated the suppression of Nrf2/Keap1 dependent antioxidant protection through induction of endoplasmic (ER) stress and Keap1 promoter DNA demethylation in human lens epithelial cells (HLECs) treated with valproic acid (VPA), an antiepileptic drug. 20 mM VPA induces ER stress and activates the unfolded protein response (UPR) within 4 h by activating the ER stress sensor proteins, such as PERK, IRE1α, and ATF6 in HLECs. Consequently, the integrated ER stress signals, such as eIF2α, ATF4, BiP, and CHOP are altered accordingly to induce ER-Ca2+ release, reactive oxygen species (ROS) overproduction, and cell death in HLECs treated with VPA. VPA also suppresses the Nrf2, catalase, and glutathione reductase expressions with significant increases in Keap1 protein. Bisulphite genomic DNA sequencing reveals the promoter DNA demethylation in the Keap1 promoter, which results in the overexpression of Keap1 mRNA and protein in HLECs treated with 20 mM VPA. VPA also alters the expression profiles of passive DNA demethylation pathway enzymes such Dnmt1, Dnmt3a, Dnmt3b, and active DNA demethylation pathway enzyme, TET1 leading to DNA demethylation in the Keap1 promoter of HLECs. Overexpressed Keap1 decreases the Nrf2 level, thereby abolishing the Nrf2 dependent antioxidant protection. This might be responsible for lenticular proteins oxidation and cataract formation.
Product Description
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Valproic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propylpentanoic acid
Valproic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®