Merck
  • CD36-mediated hematoma absorption following intracerebral hemorrhage: negative regulation by TLR4 signaling.
CD36-mediated hematoma absorption following intracerebral hemorrhage: negative regulation by TLR4 signaling.
Journal of immunology (Baltimore, Md. : 1950) (2014-05-09)
Huang Fang, Jing Chen, Sen Lin, PengFei Wang, YanChun Wang, XiaoYi Xiong, QingWu Yang
ABSTRACT
Promoting hematoma absorption is a novel therapeutic strategy for intracerebral hemorrhage (ICH); however, the mechanism of hematoma absorption is unclear. The present study explored the function and potential mechanism of CD36 in hematoma absorption using in vitro and in vivo ICH models. Hematoma absorption in CD36-deficient ICH patients was examined. Compared with patients with normal CD36 expression, CD36-deficient ICH patients had slower hematoma adsorption and aggravated neurologic deficits. CD36 expression in perihematomal tissues in wild-type mice following ICH was increased, whereas the hematoma absorption in CD36(-/-) mice was decreased. CD36(-/-) mice also showed aggravated neurologic deficits and increased TNF-α and IL-1β expression levels. The phagocytic capacity of CD36(-/-) microglia for RBCs was also decreased. Additionally, the CD36 expression in the perihematoma area after ICH in TLR4(-/-) and MyD88(-/-) mice was significantly increased, and hematoma absorption was significantly promoted, which was significantly inhibited by an anti-CD36 Ab. In vitro, TNF-α and IL-1β significantly inhibited the microglia expression of CD36 and reduced the microglia phagocytosis of RBCs. Finally, the TLR4 inhibitor TAK-242 upregulated CD36 expression in microglia, promoted hematoma absorption, increased catalase expression, and decreased the H2O2 content. These results suggested that CD36 mediated hematoma absorption after ICH, and TLR4 signaling inhibited CD36 expression to slow hematoma absorption. TLR4 inhibition could promote hematoma absorption and significantly improve neurologic deficits following ICH.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)