Merck
  • Cross-modulation and molecular interaction at the Cav3.3 protein between the endogenous lipids and the T-type calcium channel antagonist TTA-A2.
Cross-modulation and molecular interaction at the Cav3.3 protein between the endogenous lipids and the T-type calcium channel antagonist TTA-A2.
Molecular pharmacology (2013-11-12)
Magali Cazade, Cindy E Nuss, Isabelle Bidaud, John J Renger, Victor N Uebele, Philippe Lory, Jean Chemin
ABSTRACT
T-type calcium channels (T/Ca(v)3-channels) are implicated in various physiologic and pathophysiologic processes such as epilepsy, sleep disorders, hypertension, and cancer. T-channels are the target of endogenous signaling lipids including the endocannabinoid anandamide, the ω3-fatty acids, and the lipoamino-acids. However, the precise molecular mechanism by which these molecules inhibit T-current is unknown. In this study, we provided a detailed electrophysiologic and pharmacologic analysis indicating that the effects of the major N-acyl derivatives on the Ca(v)3.3 current share many similarities with those of TTA-A2 [(R)-2-(4-cyclopropylphenyl)-N-(1-(5-(2,2,2-trifluoroethoxy)pyridin-2-yl)ethyl)acetamide], a synthetic T-channel inhibitor. Using radioactive binding assays with the TTA-A2 derivative [(3)H]TTA-A1 [(R)-2-(4-(tert-butyl)phenyl)-N-(1-(5-methoxypyridin-2-yl)ethyl)acetamide], we demonstrated that polyunsaturated lipids, which inhibit the Ca(v)3.3 current, as NAGly (N-arachidonoyl glycine), NASer (N-arachidonoyl-l-serine), anandamide, NADA (N-arachidonoyl dopamine), NATau (N-arachidonoyl taurine), and NA-5HT (N-arachidonoyl serotonin), all displaced [(3)H]TTA-A1 binding to membranes prepared from cells expressing Ca(v)3.3, with Ki in a micromolar or submicromolar range. In contrast, lipids with a saturated alkyl chain, as N-arachidoyl glycine and N-arachidoyl ethanolamine, which did not inhibit the Ca(v)3.3 current, had no effect on [(3)H]TTA-A1 binding. Accordingly, bio-active lipids occluded TTA-A2 effect on Ca(v)3.3 current. In addition, TTA-Q4 [(S)-4-(6-chloro-4-cyclopropyl-3-(2,2-difluoroethyl)-2-oxo-1,2,3,4-tetrahydroquinazolin-4-yl)benzonitrile], a positive allosteric modulator of [(3)H]TTA-A1 binding and TTA-A2 functional inhibition, acted in a synergistic manner to increase lipid-induced inhibition of the Ca(v)3.3 current. Overall, our results demonstrate a common molecular mechanism for the synthetic T-channel inhibitors and the endogenous lipids, and indicate that TTA-A2 and TTA-Q4 could be important pharmacologic tools to dissect the involvement of T-current in the physiologic effects of endogenous lipids.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
SAFC
Glycine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dopamine hydrochloride
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, 99%, FCC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Dopamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dopamine hydrochloride solution, 1.0 mg/mL in methanol with 5% 1 M HCl (as free base), ampule of 1 mL, certified reference material, Cerilliant®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycine 1 M solution