Merck
  • [Utilization of different carbon sources by soil and endophytic strains of Penicillium funiculosum Thom].
[Utilization of different carbon sources by soil and endophytic strains of Penicillium funiculosum Thom].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2013-07-23)
I N Kurchenko, A I Vasilevskaia, L V Artyshkova, L T Nakonechnaia, E M Iur'eva
ABSTRACT
The ability of soil and endophytic strains of P. funiculosum to accumulate biomass under cultivation conditions in media containing carbon sources from mono- to polysaccharides has been studied. It has been shown that the most favorable carbon sources for the studied strains were maltose, sucrose, xylose, fructose, pectin, less assimilated lactose, arabinose, and especially microcrystalline cellulose. Significant differences in the level of biomass accumulation between soil and endophytic strains of this species in most cases were not established.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Supelco
Activated charcoal, powder
Millipore
Activated charcoal, suitable for GC
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, acid-washed with phosphoric and sulfuric acids
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm