Merck
  • Acidification due to microbial dechlorination near a trichloroethene DNAPL is overcome with pH buffer or formate as electron donor: experimental demonstration in diffusion-cells.
Acidification due to microbial dechlorination near a trichloroethene DNAPL is overcome with pH buffer or formate as electron donor: experimental demonstration in diffusion-cells.
Journal of contaminant hydrology (2013-03-19)
Jo Philips, Nele Maes, Dirk Springael, Erik Smolders
ABSTRACT
Acidification due to microbial dechlorination of trichloroethene (TCE) can limit the bio-enhanced dissolution of TCE dense non-aqueous phase liquid (DNAPL). This study related the dissolution enhancement of a TCE DNAPL to the pH buffer capacity of the medium and the type of electron donor used. In batch systems, dechlorination was optimal at pH7.1-7.5, but was completely inhibited below pH6.2. In addition, dechlorination in batch systems led to a smaller pH decrease at an increasing pH buffer capacity or with the use of formate instead of lactate as electron donor. Subsequently, bio-enhanced TCE DNAPL dissolution was quantified in diffusion-cells with a 5.5 cm central sand layer, separating a TCE DNAPL layer from an aqueous top layer. Three different pH buffer capacities (2.9 mM-17.9 mM MOPS) and lactate or formate as electron donor were applied. In the lactate fed diffusion-cells, the DNAPL dissolution enhancement factor increased from 1.5 to 2.2 with an increase of the pH buffer capacity. In contrast, in the formate fed diffusion-cells, the DNAPL dissolution enhancement factor (2.4±0.3) was unaffected by the pH buffer capacity. Measurement of the pore water pH confirmed that the pH decreased less with an increased pH buffer capacity or with formate instead of lactate as electron donor. These results suggest that the significant impact of acidification on bio-enhanced DNAPL dissolution can be overcome by the amendment of a pH buffer or by applying a non acidifying electron donor like formate.
Product Description
Supelco
Trichloroethylene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Chlorine, ≥99.5%
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Supelco
Trichloroethylene, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Chlorine, puriss., ≥99.8%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Calcium formate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Trichloroethylene, ACS reagent, ≥99.5%
Sigma-Aldrich
Thallium(I) formate, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Cesium formate, 98%
Supelco
Calcium formate, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Sodium formate-13C, 99 atom % 13C
Sigma-Aldrich
Potassium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium formate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%