Merck
  • Use of multi-parameter flow cytometry as tool to monitor the impact of formic acid on Saccharomyces carlsbergensis batch ethanol fermentations.
Use of multi-parameter flow cytometry as tool to monitor the impact of formic acid on Saccharomyces carlsbergensis batch ethanol fermentations.
Applied biochemistry and biotechnology (2013-01-30)
Cláudia Freitas, Elisabete Neves, Alberto Reis, Paula C Passarinho, Teresa Lopes da Silva
ABSTRACT
The use of lignocellulosic materials as substrate for bioethanol production is considered a cost-effective approach to make the biofuel production process economically sustainable. However, lignocellulosic hydrolysis releases toxic compounds such as weak acids which inhibit microorganism growth and ethanol production. In order to understand the physiological response of Saccharomyces carlsbergensis when fermenting glucose in the presence of formic acid (HF), the yeast growth was monitored by multi-parameter flow cytometry. Cytoplasmic membrane potential decreased as the HF concentration increased and as the yeast culture reached the stationary phase. However, the proportion of cells with permeabilized membrane did not increase with the HF concentration increase. The accumulation of reactive oxygen species was also monitored. Control and fermentations at low HF concentrations (<1 g/L) resulted in a high proportion of highly oxidized cells at the stationary phase. The multi-parameter flow cytometry approach proved to be a useful tool to monitor the physiological stress response of S. carlsbergensis growth and ethanol production in the presence of HF, an inhibitor present in lignocellulosic hydrolysates. The information here obtained at near real time can be used to enhance second-generation bioethanol production process efficiency.
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Potassium formate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium formate-13C, 99 atom % 13C
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Potassium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Cesium formate, 98%
Sigma-Aldrich
Thallium(I) formate, 97%
Sigma-Aldrich
Calcium formate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Supelco
Calcium formate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland