Merck
  • Determination of phenylephrine in human plasma using ultra-performance liquid chromatography-tandem mass spectrometry.
Determination of phenylephrine in human plasma using ultra-performance liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-01-15)
Sheng Feng, Qian Zhao, Ji Jiang, Pei Hu
ABSTRACT
This paper described a sensitive and rapid method based on ultra-performance liquid chromatography coupled to tandem mass spectrometry (UPLC-MS/MS) for the determination of phenylephrine in human plasma. Plasma samples were pre-purified by solid-phase extraction (SPE). The chromatographic separation was achieved with BEH HILIC column using a mixture of 10mM pH 3.5 ammonium formate and acetonitrile (10:90, v/v) under isocratic conditions at a flow rate of 0.4 mL/min. The mass spectrometry was carried out using positive electrospray ionization (ESI) and data acquisition was carried out in the multiple reaction monitoring (MRM) mode. The method was fully validated over the concentration range of 10.0-5000 pg/mL. The lower limit of quantification (LLOQ) was 10.0 pg/mL. Inter- and intra-batch precision was less than 15% and the accuracy was within 85-115%. Extraction recovery was 78.5%. Selectivity, matrix effects and stability were also validated. The method was applied to the pharmacokinetic study of phenylephrine hydrochloride in Chinese subjects.
Product Description
Supelco
Calcium formate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Calcium formate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Cesium formate, 98%
Sigma-Aldrich
Thallium(I) formate, 97%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Potassium formate, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Sodium formate-13C, 99 atom % 13C
Sigma-Aldrich
Potassium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%