Merck
  • Synthesis of (-)-swainsonine and (-)-8-epi-swainsonine by the addition of allenylmetals to chiral α,β-alkoxy sulfinylimines.
Synthesis of (-)-swainsonine and (-)-8-epi-swainsonine by the addition of allenylmetals to chiral α,β-alkoxy sulfinylimines.
Organic letters (2011-11-16)
Julien Louvel, Fabrice Chemla, Emmanuel Demont, Franck Ferreira, Alejandro Pérez-Luna
ABSTRACT
The asymmetric synthesis of (-)-swainsonine and (-)-8-epi-swainsonine is reported through the addition of either the allenylzinc or the allenyl lithio cyanocuprate reagents derived from [3-(methoxymethoxy)prop-1-ynyl]trimethylsilane to enantiopure α,β-dialkoxy N-tert-butanesulfinylimines derived from d-erythronolactone.
Sigma-Aldrich
Swainsonine, from Metarrhizium anisopliae, ≥98% (TLC)
Sigma-Aldrich
Swainsonine, synthetic