Merck
  • A comparative study of the responses of lithium borate and calcium sulphate phosphors in a TL personal dosemeter.
A comparative study of the responses of lithium borate and calcium sulphate phosphors in a TL personal dosemeter.
Radiation protection dosimetry (2011-02-08)
M Ceretti, G Costa, S Romani, F Bobba
ABSTRACT
This work summarises the results of a series of experiences made on Panasonic UD-802AS, a multi-element dosemeter that is currently used in Caorso Nuclear Power Plant for personnel external monitoring. Two main topics have been considered: energy response and the influence of working place dose rate on relative readings of the elements at the end of the monitoring period. The presence of four elements of different materials and filtrations gives the possibility of evaluating radiation energy by a simple algorithm, so that an accurate estimate of the personal dose equivalent can be provided; the good results of the participation of Caorso Individual Monitoring Service (IMS) to 'EURADOS 2008 intercomparison on whole-body dosemeters' account for the reliability of the method. A study on the response in low-dose rate fields is also reported, giving in practice interesting information about operational conditions.
Sigma-Aldrich
Lithium metaborate, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lithium metaborate, 99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lithium metaborate, ACS reagent, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lithium tetraborate, ≥99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lithium tetraborate, ≥99.9% trace metals basis