Skip to Content
Merck
All Photos(3)

Key Documents

222534

Sigma-Aldrich

Lithium tetraborate

≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2B4O7
CAS Number:
Molecular Weight:
169.12
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

≥99.9% trace metals basis

form

powder

mp

760-880 °C (lit.)

density

≥0.25 g/mL at 25 °C (lit.)

SMILES string

[Li+].[Li+].[O-]B(OB=O)OB([O-])OB=O

InChI

1S/B4O7.2Li/c5-1-9-3(7)11-4(8)10-2-6;;/q-2;2*+1

InChI key

PSHMSSXLYVAENJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium tetraborate (Li2B4O7) is a piezoelectric material. It can be synthesized from boric acid. Its pyroelectric, piezoelectric and dielectric properties have been evaluated. It is a widely used material in the field of non-linear optics. Its electronic structure has been investigated by x-ray photoelectron spectroscopy (XPS) and quantum mechanical modelling methods.
Lithium tetraborate is a pyroelectric and piezoelectric material with potential applications in pressure sensors and surface acoustic wave (SAW) devices. In the presence of activators such as terbium and cupric ions, it shows thermoluminesence property.

Application

Used as a spectroscopic fluxing agent.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elastic, piezoelectric, acousto-optic and electro-optic properties of Li2B4O7.
Shiosaki T, et al.
Japanese Journal of Applied Physics, 24(S1), 25-25 (1985)
Pyroelectric and piezoelectric properties of lithium tetraborate single crystal.
Bhalla AS, et al.
Japanese Journal of Applied Physics, 24(S2), 727-727 (1985)
Thermoluminescence in lithium tetraborate doped with activators.
Kutomi Y and Takeuchi N.
Journal of Materials Science Letters, 5(1), 51-53 (1986)
Pyroelectric and piezoelectric properties of lithium tetraborate single crystal.
Bhalla AS, et al.
Jpn J. App. Phys., Part I, 24(S2), 727-727 (1985)
Lithium isotope analysis by thermal ionization mass spectrometry of lithium tetraborate.
Chan LH.
Analytical Chemistry, 59(22), 2662-2665 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service