Merck
  • Chiral aminonaphthol-catalyzed enantioselective carbonyl addition of diethylzinc to aromatic aldehydes high-throughput screened by CD-HPLC analysis.
Chiral aminonaphthol-catalyzed enantioselective carbonyl addition of diethylzinc to aromatic aldehydes high-throughput screened by CD-HPLC analysis.
Chirality (2005-08-17)
Ling Xu, Xiumin Shen, Cong Zhang, Koichi Mikami
ABSTRACT
Optically active aminonaphthols derivatives are obtained by condensation of 2-naphthol, substituted benzaldehyde, and (S)-methylbenzylamine under mild conditions, without side products. Their absolute configurations are determined by X-ray crystallographic analysis. The addition of diethylzinc to aromatic aldehydes is considerably accelerated by the presence of a catalytic amount of crystalline to give, after hydrolysis, the corresponding 1-phenylpropanol in good enantiomeric purity, as determined by CD-HPLC analysis as HTPS (high-throughput screening).
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes
Sigma-Aldrich
Diethylzinc, ≥52 wt. % Zn basis
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene