Merck
  • Light-dependent inhibition of clathrin-mediated endocytosis in yeast unveils conserved functions of the AP2 complex.
Light-dependent inhibition of clathrin-mediated endocytosis in yeast unveils conserved functions of the AP2 complex.
iScience (2023-09-28)
Davia Prischich, Núria Camarero, Javier Encinar Del Dedo, Maria Cambra-Pellejà, Judit Prat, Laura Nevola, Andrés Martín-Quirós, Elena Rebollo, Laura Pastor, Ernest Giralt, María Isabel Geli, Pau Gorostiza
ABSTRACT
Clathrin-mediated endocytosis (CME) is an essential cellular process, conserved among eukaryotes. Yeast constitutes a powerful genetic model to dissect the complex endocytic machinery, yet there is a lack of specific pharmacological agents to interfere with CME in these organisms. TL2 is a light-regulated peptide inhibitor targeting the AP2-β-adaptin/β-arrestin interaction and that can photocontrol CME with high spatiotemporal precision in mammalian cells. Here, we study endocytic protein dynamics by live-cell imaging of the fluorescently tagged coat-associated protein Sla1-GFP, demonstrating that TL2 retains its inhibitory activity in S. cerevisiae spheroplasts. This is despite the β-adaptin/β-arrestin interaction not being conserved in yeast. Our data indicate that the AP2 α-adaptin is the functional target of activated TL2. We identified as interacting partners for the α-appendage, the Eps15 and epsin homologues Ede1 and Ent1. This demonstrates that endocytic cargo loading and sensing can be executed by conserved molecular interfaces, regardless of the proteins involved.
Product Description
Roche
Antipain dihydrochloride, crystalline, suitable for cell culture, pkg of 10 mg
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1