Merck
  • SARS-CoV-2 and Its Bacterial Co- or Super-Infections Synergize to Trigger COVID-19 Autoimmune Cardiopathies.
SARS-CoV-2 and Its Bacterial Co- or Super-Infections Synergize to Trigger COVID-19 Autoimmune Cardiopathies.
International journal of molecular sciences (2023-08-12)
Robert Root-Bernstein, Jack Huber, Alison Ziehl, Maja Pietrowicz
ABSTRACT
Autoimmune cardiopathies (AC) following COVID-19 and vaccination against SARS-CoV-2 occur at significant rates but are of unknown etiology. This study investigated the possible roles of viral and bacterial mimicry, as well as viral-bacterial co-infections, as possible inducers of COVID-19 AC using proteomic methods and enzyme-linked immunoadsorption assays. BLAST and LALIGN results of this study demonstrate that SARS-CoV-2 shares a significantly greater number of high quality similarities to some cardiac protein compared with other viruses; that bacteria such as Streptococci, Staphylococci and Enterococci also display very significant similarities to cardiac proteins but to a different set than SARS-CoV-2; that the importance of these similarities is largely validated by ELISA experiments demonstrating that polyclonal antibodies against SARS-CoV-2 and COVID-19-associated bacteria recognize cardiac proteins with high affinity; that to account for the range of cardiac proteins targeted by autoantibodies in COVID-19-associated autoimmune myocarditis, both viral and bacterial triggers are probably required; that the targets of the viral and bacterial antibodies are often molecularly complementary antigens such as actin and myosin, laminin and collagen, or creatine kinase and pyruvate kinase, that are known to bind to each other; and that the corresponding viral and bacterial antibodies recognizing these complementary antigens also bind to each other with high affinity as if they have an idiotype-anti-idiotype relationship. These results suggest that AC results from SARS-CoV-2 infections or vaccination complicated by bacterial infections. Vaccination against some of these bacterial infections, such as Streptococci and Haemophilus, may therefore decrease AC risk, as may the appropriate and timely use of antibiotics among COVID-19 patients and careful screening of vaccinees for signs of infection such as fever, diarrhea, infected wounds, gum disease, etc.
Sigma-Aldrich
Anti-Coxsackievirus B Blend Antibody, clone Blend of 6s, ascites fluid, Chemicon®
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Rabbit Anti-Goat IgG Antibody, HRP conjugate, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Anti-Adenovirus Antibody, Chemicon®, from goat