Merck
  • Active DNA damage eviction by HLTF stimulates nucleotide excision repair.
Active DNA damage eviction by HLTF stimulates nucleotide excision repair.
Molecular cell (2022-03-11)
Marvin van Toorn, Yasemin Turkyilmaz, Sueji Han, Di Zhou, Hyun-Suk Kim, Irene Salas-Armenteros, Mihyun Kim, Masaki Akita, Franziska Wienholz, Anja Raams, Eunjin Ryu, Sukhyun Kang, Arjan F Theil, Karel Bezstarosti, Maria Tresini, Giuseppina Giglia-Mari, Jeroen A Demmers, Orlando D Schärer, Jun-Hyuk Choi, Wim Vermeulen, Jurgen A Marteijn
ABSTRACT
Nucleotide excision repair (NER) counteracts the onset of cancer and aging by removing helix-distorting DNA lesions via a "cut-and-patch"-type reaction. The regulatory mechanisms that drive NER through its successive damage recognition, verification, incision, and gap restoration reaction steps remain elusive. Here, we show that the RAD5-related translocase HLTF facilitates repair through active eviction of incised damaged DNA together with associated repair proteins. Our data show a dual-incision-dependent recruitment of HLTF to the NER incision complex, which is mediated by HLTF's HIRAN domain that binds 3'-OH single-stranded DNA ends. HLTF's translocase motor subsequently promotes the dissociation of the stably damage-bound incision complex together with the incised oligonucleotide, allowing for an efficient PCNA loading and initiation of repair synthesis. Our findings uncover HLTF as an important NER factor that actively evicts DNA damage, thereby providing additional quality control by coordinating the transition between the excision and DNA synthesis steps to safeguard genome integrity.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Spironolactone, 97.0-103.0%
Sigma-Aldrich
Anti-Goat IgG (H+L), highly cross-adsorbed, CF770 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (H+L), highly cross-adsorbed, CF 770 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Anti-Rat IgG (H+L), highly cross-adsorbed, CF770 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (H+L), highly cross-adsorbed, CF 680 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Roche
Anti-HA-Biotin, High Affinity (3F10), from rat IgG1
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 680 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 770 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Millipore
Benzonase® Nuclease, Purity > 99%, Effective viscosity reduction and removal of nucleic acids from protein solutions
Sigma-Aldrich
DAPI, for nucleic acid staining