Merck
  • Rescue of α-synuclein aggregation in Parkinson's patient neurons by synergistic enhancement of ER proteostasis and protein trafficking.
Rescue of α-synuclein aggregation in Parkinson's patient neurons by synergistic enhancement of ER proteostasis and protein trafficking.
Neuron (2021-11-19)
Iva Stojkovska, Willayat Y Wani, Friederike Zunke, Nandkishore R Belur, Egor A Pavlenko, Nkatha Mwenda, Karan Sharma, Laetitia Francelle, Joseph R Mazzulli
ABSTRACT
Neurodegenerative disorders are characterized by a collapse in proteostasis, as shown by the accumulation of insoluble protein aggregates in the brain. Proteostasis involves a balance of protein synthesis, folding, trafficking, and degradation, but how aggregates perturb these pathways is unknown. Using Parkinson's disease (PD) patient midbrain cultures, we find that aggregated α-synuclein induces endoplasmic reticulum (ER) fragmentation and compromises ER protein folding capacity, leading to misfolding and aggregation of immature lysosomal β-glucocerebrosidase. Despite this, PD neurons fail to initiate the unfolded protein response, indicating perturbations in sensing or transducing protein misfolding signals in the ER. Small molecule enhancement of ER proteostasis machinery promotes β-glucocerebrosidase solubility, while simultaneous enhancement of trafficking improves ER morphology, lysosomal function, and reduces α-synuclein. Our studies suggest that aggregated α-synuclein perturbs the ability of neurons to respond to misfolded proteins in the ER, and that synergistic enhancement of multiple proteostasis branches may provide therapeutic benefit in PD.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Anti-Glucocerebrosidase (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-α-Synuclein antibody, Mouse monoclonal, clone Syn211, purified from hybridoma cell culture
Roche
Bovine Serum Albumin Fraction V, heat shock, fatty acid free, from bovine serum
Sigma-Aldrich
Dantrolene sodium salt
Sigma-Aldrich
(+)-cis-Diltiazem hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
Monoclonal Anti-Cathepsin D antibody produced in mouse, clone CTD-19, ascites fluid
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Conduritol B Epoxide, Conduritol B Epoxide, CAS 6090-95-5, is an irreversible Inhibitor of glucocerebrosidase in neurons. Also inhibits α-glucosidase activity in a variety of species.
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial