Merck
  • p53 directs leader cell behavior, migration, and clearance during epithelial repair.
p53 directs leader cell behavior, migration, and clearance during epithelial repair.
Science (New York, N.Y.) (2022-02-11)
Kasia Kozyrska, Giulia Pilia, Medhavi Vishwakarma, Laura Wagstaff, Maja Goschorska, Silvia Cirillo, Saad Mohamad, Kelli Gallacher, Rafael E Carazo Salas, Eugenia Piddini
ABSTRACT
Epithelial cells migrate across wounds to repair injured tissue. Leader cells at the front of migrating sheets often drive this process. However, it is unclear how leaders emerge from an apparently homogeneous epithelial cell population. We characterized leaders emerging from epithelial monolayers in cell culture and found that they activated the stress sensor p53, which was sufficient to initiate leader cell behavior. p53 activated the cell cycle inhibitor p21WAF1/CIP1, which in turn induced leader behavior through inhibition of cyclin-dependent kinase activity. p53 also induced crowding hypersensitivity in leader cells such that, upon epithelial closure, they were eliminated by cell competition. Thus, mechanically induced p53 directs emergence of a transient population of leader cells that drive migration and ensures their clearance upon epithelial repair.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
RO-3306, ≥98% (HPLC)
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
(−)-Blebbistatin, solid, synthetic
Sigma-Aldrich
Dipeptidyl Peptidase III human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
Rac1 Inhibitor II, Z62954982, InSolution, ≥93%