MilliporeSigma
  • A Genetic modification that reduces ON-bipolar cells in hESC-derived retinas enhances functional integration after transplantation.
A Genetic modification that reduces ON-bipolar cells in hESC-derived retinas enhances functional integration after transplantation.
iScience (2022-01-14)
Suguru Yamasaki, Hung-Ya Tu, Take Matsuyama, Matsuri Horiuchi, Tomoyo Hashiguchi, Junki Sho, Atsushi Kuwahara, Akiyoshi Kishino, Toru Kimura, Masayo Takahashi, Michiko Mandai
ABSTRACT
Pluripotent stem cell (PSC)-derived retinal sheet transplanted in vivo can form structured photoreceptor layers, contact with host bipolar cells, and transmit light signals to host retinas. However, a major concern is the presence of graft bipolar cells that may impede host-graft interaction. In this study, we used human ESC-retinas with the deletion of Islet-1 (ISL1) gene to achieve the reduced graft ON-bipolar cells after xenotransplantation into end-stage retinal degeneration model rats. Compared with wild-type graft, ISL1 -/- hESC-retinas showed better host-graft contact, with indication of host-graft synapse formation and significant restoration of light responsiveness in host ganglion cells. We further analyzed to find out that improved functional integration of ISL1 -/- hESC-retinas seemed attributed by a better host-graft contact and a better preservation of host inner retina. ISL1 -/- hESC-retinas are promising for the efficient reconstruction of a degenerated retinal network in future clinical application.
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Rhodopsin Antibody, clone 4D2, clone 4D2, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Dihydropyridine-sensitive Calcium Channel α 1 Subunit Antibody, clone 1a, ascites fluid, clone mAB 1a, Chemicon®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-G Protein Goα Antibody, clone 2A, clone 2A, Chemicon®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Opsin Antibody, Red/Green, Chemicon®, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-NANOG Antibody, clone 7F7.1, clone 7F7.1, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Versican Antibody, clone 8D8.1, clone 8D8.1, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Peripherin-2 Antibody, clone 6B10.1, clone 6B10.1, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Glutamine Synthetase Antibody, clone GS-6, clone GS-6, Chemicon®, from mouse