Merck
  • Abnormal dopamine receptor signaling allows selective therapeutic targeting of neoplastic progenitors in AML patients.
Abnormal dopamine receptor signaling allows selective therapeutic targeting of neoplastic progenitors in AML patients.
Cell reports. Medicine (2021-03-06)
Lili Aslostovar, Allison L Boyd, Yannick D Benoit, Justin Di Lu, Juan Luis Garcia Rodriguez, Mio Nakanishi, Deanna P Porras, Jennifer C Reid, Ryan R Mitchell, Brian Leber, Anargyros Xenocostas, Ronan Foley, Mickie Bhatia
ABSTRACT
The aberrant expression of dopamine receptors (DRDs) in acute myeloid leukemia (AML) cells has encouraged the repurposing of DRD antagonists such as thioridazine (TDZ) as anti-leukemic agents. Here, we access patient cells from a Phase I dose escalation trial to resolve the cellular and molecular bases of response to TDZ, and we extend these findings to an additional independent cohort of AML patient samples tested preclinically. We reveal that in DRD2+ AML patients, DRD signaling in leukemic progenitors provides leukemia-exclusive networks of sensitivity that spare healthy hematopoiesis. AML progenitor cell suppression can be increased by the isolation of the positive enantiomer from the racemic TDZ mixture (TDZ+), and this is accompanied by reduced cardiac liability. Our study indicates that the development of DRD-directed therapies provides a targeting strategy for a subset of AML patients and potentially other cancers that acquire DRD expression upon transformation from healthy tissue.
Product Description
Supelco
N-Desmethylcitalopram hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Anti-phospho-CREB (Ser133) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Dopamine D₁ Receptor Rabbit pAb, lyophilized, Calbiochem®
Sigma-Aldrich
Anti-Dopamine D₂ Receptor Rabbit pAb, lyophilized, Calbiochem®
Supelco
Diethylpropion-D10 hydrochloride solution, 100 μg/mL in methanol (with 1% 1 M HCl, (as free base)), certified reference material, ampule of 1 mL, Cerilliant®
Sigma-Aldrich
Domperidone, powder, ≥98% (HPLC)
Sigma-Aldrich
R(+)-SCH-23390 hydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
Thioridazine hydrochloride, ≥99%
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-Dopamine D2 Receptor Antibody, Chemicon®, from rabbit
Sigma-Aldrich
(±)-SKF-38393 hydrochloride, crystalline, ≥98% (HPLC)