Merck
  • S-acylation controls SARS-CoV-2 membrane lipid organization and enhances infectivity.
S-acylation controls SARS-CoV-2 membrane lipid organization and enhances infectivity.
Developmental cell (2021-10-03)
Francisco S Mesquita, Laurence Abrami, Oksana Sergeeva, Priscilla Turelli, Enya Qing, Béatrice Kunz, Charlène Raclot, Jonathan Paz Montoya, Luciano A Abriata, Tom Gallagher, Matteo Dal Peraro, Didier Trono, Giovanni D'Angelo, F Gisou van der Goot
ABSTRACT
SARS-CoV-2 virions are surrounded by a lipid bilayer that contains membrane proteins such as spike, responsible for target-cell binding and virus fusion. We found that during SARS-CoV-2 infection, spike becomes lipid modified, through the sequential action of the S-acyltransferases ZDHHC20 and 9. Particularly striking is the rapid acylation of spike on 10 cytosolic cysteines within the ER and Golgi. Using a combination of computational, lipidomics, and biochemical approaches, we show that this massive lipidation controls spike biogenesis and degradation, and drives the formation of localized ordered cholesterol and sphingolipid-rich lipid nanodomains in the early Golgi, where viral budding occurs. Finally, S-acylation of spike allows the formation of viruses with enhanced fusion capacity. Our study points toward S-acylating enzymes and lipid biosynthesis enzymes as novel therapeutic anti-viral targets.
Sigma-Aldrich
Anti-ZDHHC20 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-ZDHHC9 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Methyl-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Filipin III from Streptomyces filipinensis, ≥85% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
FTY720, ≥98% (HPLC)
Sigma-Aldrich
APT1 Inhibitor, palmostatin B, The APT1 Inhibitor, palmostatin B controls the biological activity of APT1.
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Hydroxylamine hydrochloride, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Anti-zDHHC8 Antibody, from rabbit, purified by affinity chromatography
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1