Merck
  • IL1β Promotes Immune Suppression in the Tumor Microenvironment Independent of the Inflammasome and Gasdermin D.
IL1β Promotes Immune Suppression in the Tumor Microenvironment Independent of the Inflammasome and Gasdermin D.
Cancer immunology research (2020-12-29)
Máté Kiss, Lieselotte Vande Walle, Pedro H V Saavedra, Els Lebegge, Helena Van Damme, Aleksandar Murgaski, Junbin Qian, Manuel Ehling, Samantha Pretto, Evangelia Bolli, Jiri Keirsse, Pauline M R Bardet, Sana M Arnouk, Yvon Elkrim, Maryse Schmoetten, Jan Brughmans, Ayla Debraekeleer, Amelie Fossoul, Louis Boon, Geert Raes, Geert van Loo, Diether Lambrechts, Massimiliano Mazzone, Alain Beschin, Andy Wullaert, Mohamed Lamkanfi, Jo A Van Ginderachter, Damya Laoui
ABSTRACT
IL1β is a central mediator of inflammation. Secretion of IL1β typically requires proteolytic maturation by the inflammasome and formation of membrane pores by gasdermin D (GSDMD). Emerging evidence suggests an important role for IL1β in promoting cancer progression in patients, but the underlying mechanisms are ill-defined. Here, we have shown a key role for IL1β in driving tumor progression in two distinct mouse tumor models. Notably, activation of the inflammasome, caspase-8, as well as the pore-forming proteins GSDMD and mixed lineage kinase domain-like protein in the host were dispensable for the release of intratumoral bioactive IL1β. Inflammasome-independent IL1β release promoted systemic neutrophil expansion and fostered accumulation of T-cell-suppressive neutrophils in the tumor. Moreover, IL1β was essential for neutrophil infiltration triggered by antiangiogenic therapy, thereby contributing to treatment-induced immunosuppression. Deletion of IL1β allowed intratumoral accumulation of CD8+ effector T cells that subsequently activated tumor-associated macrophages. Depletion of either CD8+ T cells or macrophages abolished tumor growth inhibition in IL1β-deficient mice, demonstrating a crucial role for CD8+ T-cell-macrophage cross-talk in the antitumor immune response. Overall, these results support a tumor-promoting role for IL1β through establishing an immunosuppressive microenvironment and show that inflammasome activation is not essential for release of this cytokine in tumors.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Nω-Hydroxy-nor-L-arginine, Diacetate Salt, A potent, selective, competitive, and high affinity inhibitor of arginase.
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, ≥3,000 units/mg protein, BioReagent, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder