Merck
  • Ionic Polyureas-A Novel Subclass of Poly(Ionic Liquid)s for CO2 Capture.
Ionic Polyureas-A Novel Subclass of Poly(Ionic Liquid)s for CO2 Capture.
Membranes (2020-09-24)
Sofia M Morozova, Elena I Lozinskaya, Haritz Sardon, Fabian Suárez-García, Petr S Vlasov, Régis Vaudemont, Yakov S Vygodskii, Alexander S Shaplov
ABSTRACT
The growing concern for climate change and global warming has given rise to investigations in various research fields, including one particular area dedicated to the creation of solid sorbents for efficient CO2 capture. In this work, a new family of poly(ionic liquid)s (PILs) comprising cationic polyureas (PURs) with tetrafluoroborate (BF4) anions has been synthesized. Condensation of various diisocyanates with novel ionic diamines and subsequent ion metathesis reaction resulted in high molar mass ionic PURs (Mw = 12 ÷ 173 × 103 g/mol) with high thermal stability (up to 260 °C), glass transition temperatures in the range of 153-286 °C and remarkable CO2 capture (10.5-24.8 mg/g at 0 °C and 1 bar). The CO2 sorption was found to be dependent on the nature of the cation and structure of the diisocyanate. The highest sorption was demonstrated by tetrafluoroborate PUR based on 4,4'-methylene-bis(cyclohexyl isocyanate) diisocyanate and aromatic diamine bearing quinuclidinium cation (24.8 mg/g at 0 °C and 1 bar). It is hoped that the present study will inspire novel design strategies for improving the sorption properties of PILs and the creation of novel effective CO2 sorbents.
Sigma-Aldrich
1,5-Dibromopentane, 97%
Sigma-Aldrich
Tin(II) 2-ethylhexanoate, 92.5-100.0%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
3,3′-Diamino-N-methyldipropylamine, 96%
Sigma-Aldrich
Iodoethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Sigma-Aldrich
3-Aminoquinuclidine dihydrochloride, 98%