Skip to Content
Merck
All Photos(3)

Key Documents

188441

Sigma-Aldrich

3,3′-Diamino-N-methyldipropylamine

96%

Synonym(s):

(7-Amino-4-methyl-4-azaheptyl)amine, 1,7-Diamino-4-aza(methyl)heptane, 1,7-Diamino-4-methyl-4-azaheptane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3N(CH2CH2CH2NH2)2
CAS Number:
Molecular Weight:
145.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

6 mmHg ( 110 °C)

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.4725 (lit.)

bp

110-112 °C/6 mmHg (lit.)

density

0.901 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CN(CCCN)CCCN

InChI

1S/C7H19N3/c1-10(6-2-4-8)7-3-5-9/h2-9H2,1H3

InChI key

KMBPCQSCMCEPMU-UHFFFAOYSA-N

Application

3,3′-Diamino-N-methyldipropylamine was used in the synthesis of:
  • new bischromone derivatives, potential anticancer drugs
  • polyamine derivatives containing dimeric quinolone, cinnoline and phthalimide moieties
  • 3-(2,4-dinitroanilino)-3′-amino-N-methyldipropylamine

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Agata Szulawska-Mroczek et al.
Archiv der Pharmazie, 346(1), 34-43 (2012-10-31)
The synthesis of new bischromone derivatives (4a-c and 5a-c) as potential anticancer drugs is described. The difference in the reactivity between 4-oxo-4H-chromene-3-carboxylic acid 2 (or its methyl ester 3) and 4-oxo-4H-chromene-3-carbonyl chloride 1 with three different polyamines: 3,3'-diamino-N-methyldipropylamine (a), 1,4-bis(3-aminopropyl)piperazine
R G Anderson et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(15), 4838-4842 (1984-08-01)
We report the synthesis of a probe that permits the visualization by electron microscopy of acidic organelles in intact cells. This probe, 3-(2,4-dinitroanilino)-3'-amino-N-methyldipropylamine (DAMP), is a basic congener of dinitrophenol that readily diffuses into intact cells. Its primary and tertiary
Marta Szumilak et al.
European journal of medicinal chemistry, 45(12), 5744-5751 (2010-10-27)
The synthesis of new polyamine derivatives containing dimeric quinoline (3a-c), cinnoline (4a-c) and phthalimide (7a-c and 8a-c) moieties is described. Three different polyamines: (1,4-bis(3-aminopropyl)piperazine (a), 4,9-dioxa-1,12-dodecanediamine (b), 3,3'-diamino-N-methyldipropylamine (c) were used as linkers. The new compounds were obtained according to
Carla Cruz et al.
PloS one, 6(11), e27078-e27078 (2011-12-02)
Useful probes of the intracellular environment that target a specific organelle in order to allow direct observation of the changes in these regions is of high current interest. Macrocyclic ligands have already revealed themselves as important selective hosts in some
Lucas Vu et al.
Theranostics, 2(12), 1160-1173 (2013-02-06)
A focused library of twenty-one cationic poly(amino ethers) was synthesized following ring-opening polymerization of two diglycidyl ethers by different oligoamines. The polymers were screened in parallel for plasmid DNA (pDNA) delivery, and transgene expression efficacies of individual polymers were compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service