Merck
  • Fibronectin/thermo-responsive polymer scaffold as a dynamic ex vivo niche for mesenchymal stem cells.
Fibronectin/thermo-responsive polymer scaffold as a dynamic ex vivo niche for mesenchymal stem cells.
Journal of materials science. Materials in medicine (2020-12-01)
Laura Ramalho, Salima Nedjari, Roberto Guarino, Firas Awaja, Dencho Gugutkov, George Altankov
ABSTRACT
In this paper, we created a dynamic adhesive environment (DAE) for adipose tissue-derived mesenchymal stem cells (ADMSCs) cultured on smart thermo-responsive substrates, i.e., poly (N-isopropyl acrylamide) (PNIPAM), via introducing periodic changes in the culture temperature. We further explored the particular role of adsorbed fibronectin (FN), an important cell adhesive protein that was recently attributed to the recruitment of stem cells in the niche. The engineered FN/PNIPAM DAE system significantly increased the symmetric renewal of ADMSCs, particularly between passages 7 and 9 (p7-p9), before it dropped down to the level of the control (FN-coated TC polystyrene). This decline in the growth curve was consistent with the increased number of senescent cells, the augmented average cell size and the suppressed FN matrix secretion at late passages (p10-p12), all of them characteristic for stem cells ageing, which equivocally tended to slow down at our DAE system. FN supported also the osteogenic response of ADMSCs (apart from the previous observations with plain PNIPAM substrata) indicated by the significant increase of alkaline phosphatase (ALP) activity at days 7 and 14. The minimal changes in the Ca deposition, however, suggest a restricted effect of DAE on the early osteogenic response of ADMSCs only. Thus, the engineering of niche-like DAE involving FN uncovers a new tissue engineering strategy for gaining larger amounts of functionally active stem cells for clinical application.
Sigma-Aldrich
Alkaline buffer solution, 1.5 M, pH 10.3 (25 °C)
Sigma-Aldrich
Alizarin Red S, certified by the Biological Stain Commission
Sigma-Aldrich
Petri dishes, polystyrene, size 60 mm × 15 mm
Sigma-Aldrich
Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Senescence Cells Histochemical Staining Kit, sufficient for 100 tests
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture