Merck
  • Hippocampal Radial Glial Subtypes and Their Neurogenic Potential in Human Fetuses and Healthy and Alzheimer's Disease Adults.
Hippocampal Radial Glial Subtypes and Their Neurogenic Potential in Human Fetuses and Healthy and Alzheimer's Disease Adults.
Cerebral cortex (New York, N.Y. : 1991) (2018-05-04)
Sara Cipriani, Isidre Ferrer, Eleonora Aronica, Gabor G Kovacs, Catherine Verney, Jeannette Nardelli, Suonavy Khung, Anne-Lise Delezoide, Ivan Milenkovic, Sowmyalakshmi Rasika, Philippe Manivet, Jean-Louis Benifla, Nicolas Deriot, Pierre Gressens, Homa Adle-Biassette
ABSTRACT
Neuropathological conditions might affect adult granulogenesis in the adult human dentate gyrus. However, radial glial cells (RGCs) have not been well characterized during human development and aging. We have previously described progenitor and neuronal layer establishment in the hippocampal pyramidal layer and dentate gyrus from embryonic life until mid-gestation. Here, we describe RGC subtypes in the hippocampus from 13 gestational weeks (GW) to mid-gestation and characterize their evolution and the dynamics of neurogenesis from mid-gestation to adulthood in normal and Alzheimer's disease (AD) subjects. In the pyramidal ventricular zone (VZ), RGC density declined with neurogenesis from mid-gestation until the perinatal period. In the dentate area, morphologic and antigenic differences among RGCs were observed from early ages of development to adulthood. Density and proliferative capacity of dentate RGCs as well as neurogenesis were strongly reduced during childhood until 5 years, few DCX+ cells are seen in adults. The dentate gyrus of both control and AD individuals showed Nestin+ and/or GFAPδ+ cells displaying different morphologies. In conclusion, pools of morphologically, antigenically, and topographically diverse neural progenitor cells are present in the human hippocampus from early developmental stages until adulthood, including in AD patients, while their neurogenic potential seems negligible in the adult.
Sigma-Aldrich
Anti-Glial fibrillary acidic protein δ Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Nestin Antibody, clone 10C2, clone 10C2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Nestin, Human Antibody, serum, from rabbit
Sigma-Aldrich
Anti-Doublecortin Antibody, serum, from guinea pig