Skip to Content
Merck
All Photos(2)

Key Documents

MAB5326

Sigma-Aldrich

Anti-Nestin Antibody, clone 10C2

clone 10C2, Chemicon®, from mouse

Synonym(s):

Anti-Nestin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

10C2, monoclonal

species reactivity

human

should not react with

rat, mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

input

sample type epithelial cells
sample type neural stem cell(s)

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Nestin, a large intermediate filament protein (class Type VI) expressed during development and in myotendinous and neuromuscular junctions. Nestin expression is restricted, typically disappearing by E18. Nestin is thought to be a reasonable neuronal marker; however, recent studies have found nestin expression in other cell types such as endothelial cells {Folia Biologica, 44(5):155, 1998}. Nestin identifies the most primitive neuroepithelium but also identifies many other embryonic tissues, so it is not specific for CNS. Nestin expression is seen in almost all GBMs (Glioblastoma mulitformes) and many melanomas (both primary and metastatic) but not in any metastatic carcinoma.

Specificity

Reacts with human Nestin, with weak reactivity to monkey nestin. Not reactive with rodent nestin.

Immunogen

Epitope: human
Fusion protein.

Application

Immunoblotting: 1:1,000-1:2,000 on denatured protein. Western blot: 1:1000-1:5000. Recognizes a 220-240kDA band on denaturing SDS gels. There is also a 60-64kDa band present in most lysates, the identity of this smaller fragment is unknown but it is not seen in lysates not containing human nestin protein.

WI-38 (ATCC# CCL75) can be used as a positive control. U251 cell line is also positive.

Immunocytochemistry: 1:200 on cells fixed in 4% paraformaldehyde and permeabilized with 0.02% Triton

Immunohistochemistry: paraformaldehyde is the preferred fixative: 1:200 on formalin fixed, paraffin embedded tissue sections. Use triton 0.02% in block only; dilution buffer 0.1% BSA; incubations 1-2 hours RT. Human fetal brain is suggested as a positive control. (CA Messam et al, Develop. Brain Res, 134:87-92, 2002; Gu, et al., Brain Res. 943:174-180, 2002). Filamentous staining in the cytoplasm of cells

Optimal working dilutions must be determined by end user.
Research Category
Stem Cell Research
Research Sub Category
Neural Stem Cells
This Anti-Nestin Antibody, clone 10C2 is validated for use in IC, IH, IH(P), WB for the detection of Nestin.

Target description

200-220 kDa

Linkage

Replaces: AB5922

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Protein A purified

Storage and Stability

Antibodies may be stored at 4°C or -20°C. Avoid repeated freeze/thaw cycles.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Analysis Note

Control
Positive Control: WI-38 (ATCC# CCL75) or U251 cell lines

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cannabinoids induce glioma stem-like cell differentiation and inhibit gliomagenesis.
Aguado, Tania, et al.
The Journal of Biological Chemistry, 282, 6854-6862 (2007)
A novel myoepithelial/progenitor cell marker in the breast?
Zdenek Kolar,Jiri Ehrmann,Gulisa Turashvili,Jan Bouchal,Jaroslav Mokry
Virchows Archiv null
C A Messam et al.
Experimental neurology, 161(2), 585-596 (2000-02-25)
The presence of the intermediate filament protein nestin has been the predominant marker used to describe stem and progenitor cells in the mammalian CNS. In this study, a 998-bp fragment in the 3' region of the nestin mRNA was cloned
Expression of stem-cell markers (cytokeratin 15 and nestin) in primary adnexal neoplasms-clues to etiopathogenesis.
Mahalingam M, Srivastava A, Hoang MP
The American Journal of Dermatopathology null
Multiple neurogenic and neurorescue effects of human mesenchymal stem cell after transplantation in an experimental model of Parkinson's disease.
Cova, Lidia, et al.
Brain Research, 1311, 12-27 (2010)

Articles

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

See All

Protocols

A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.

A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service